Skip to Content
Merck
All Photos(1)

Key Documents

8.20828

Sigma-Aldrich

1-Methyl-4-piperidinone

for synthesis

Synonym(s):

1-Methyl-4-piperidinone, N-Methyl-γ-piperidone

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€84.10

€84.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€84.10

About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
MDL number:
UNSPSC Code:
12352115
EC Index Number:
215-895-5
NACRES:
NA.22

€84.10


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

pH

12 (20 °C, 100 g/L in H2O)

bp

60-62 °C/19 hPa

transition temp

flash point 58 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1(CCC(=O)CC1)C

InChI

1S/C6H11NO/c1-7-4-2-6(8)3-5-7/h2-5H2,1H3

InChI key

HUUPVABNAQUEJW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.972 - 0.978
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service