Skip to Content
Merck
All Photos(1)

Documents

8.20819

Sigma-Aldrich

2-Methyl-2,4-pentanediol

for synthesis

Synonym(s):

2-Methyl-2,4-pentanediol, Hexylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O2
CAS Number:
Molecular Weight:
118.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-489-0
NACRES:
NA.22

vapor pressure

0.07 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

425 °C (DIN 51794)

potency

3692 mg/kg LD50, oral (Rat)
8000 mg/kg LD50, skin (Rabbit)

expl. lim.

1-9.9 % (v/v)

mp

-40 °C

transition temp

flash point 93 °C (DIN 51758)

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3

InChI key

SVTBMSDMJJWYQN-UHFFFAOYSA-N

Application


  • Pickering Emulsion Enhancement: A study explored the application of 2-Methyl-2,4-pentanediol in the formation of Pickering emulsions, showing how it enhances antioxidant and anti-aging activities when combined with basil extract, emphasizing its role in innovative cosmetic formulations (Rosdianto et al., 2023).

  • Improving Cement Hydration: The modification of alkali-activated slag by organic additives, including 2-Methyl-2,4-pentanediol was investigated, correlating the hydration process to the evolution of material properties, which has implications for construction materials technology (Bilek et al., 2023).

  • Impact on Skin Penetration: An investigation into the impact of 2-Methyl-2,4-pentanediol on stratum corneum lipids revealed its influence on the skin penetration of triamcinolone acetonide, underlining its importance in pharmaceutical formulations and dermal delivery systems (Sigg and Daniels, 2021).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.921 - 0.923
Identity (IR): passes test

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service