Skip to Content
Merck
All Photos(1)

Key Documents

8.20798

Sigma-Aldrich

2-Methylfuran

(stabilised) for synthesis

Synonym(s):

2-Methylfuran, Silvan

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€47.80
1 L
€141.00

€47.80


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€47.80
1 L
€141.00

About This Item

Empirical Formula (Hill Notation):
C5H6O
CAS Number:
Molecular Weight:
82.10
MDL number:
UNSPSC Code:
12352005
EC Index Number:
208-594-5
NACRES:
NA.22

€47.80


Estimated to ship on19 May 2025


Request a Bulk Order

vapor pressure

139 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

167 mg/kg LD50, oral (Rat)

bp

63-66 °C/1013 hPa

mp

-89 °C

transition temp

flash point -27 °C

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[o]1c(ccc1)C

InChI

1S/C5H6O/c1-5-3-2-4-6-5/h2-4H,1H3

InChI key

VQKFNUFAXTZWDK-UHFFFAOYSA-N

Application

  • Electrocatalytic reduction of furfural: A study described the selective reduction of furfural to 2-methylfuran via electrode-electrolyte interface modification, providing insights into advanced electrocatalysis methods for producing bio-based chemicals (Ji et al., 2024).
  • Hydrodeoxygenation of furfural: Research demonstrated the conversion of furfural to 2-methylfuran over a Cu-Co catalyst confined by a hollow carbon cage, showcasing enhanced charge transfer and alloy structure for efficient chemical synthesis (Dou et al., 2024).
  • Environmental genotoxicity study: An investigation into the genotoxic effects of 2-methyl furan on male Sprague-Dawley rats over a 120-day exposure period highlighted the potential environmental and health risks associated with this compound (Cao et al., 2024).
  • Cytotoxicity trend prediction: A study explored the mechanism-based cytotoxicity trends of furan-containing pollutants, including 2-methylfuran, when present in mixtures, aiding in the prediction and management of environmental pollution risks (Li et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.913 - 0.916
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service