Skip to Content
Merck
All Photos(1)

Key Documents

8.20789

Sigma-Aldrich

Methylcyclopentane

for synthesis

Synonym(s):

Methylcyclopentane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€31.50
250 ML
€94.70

€31.50


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€31.50
250 ML
€94.70

About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
MDL number:
UNSPSC Code:
12352001
EC Index Number:
202-503-2
NACRES:
NA.22

€31.50


Estimated to ship on19 May 2025


Request a Bulk Order

vapor pressure

150 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

form

liquid

autoignition temp.

315 °C

bp

72 °C/1013 mbar

mp

-142 °C

transition temp

flash point -17 °C

density

0.74 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1(CCCC1)C

InChI

1S/C6H12/c1-6-4-2-3-5-6/h6H,2-5H2,1H3

InChI key

GDOPTJXRTPNYNR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.746 - 0.749
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service