Skip to Content
Merck
All Photos(1)

Key Documents

8.20747

Sigma-Aldrich

Potassium borohydride

for synthesis

Synonym(s):

Potassium borohydride, Potassium tetrahydroborate, Potassium boranate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€53.60
100 G
€109.00
1 KG
€818.00

€53.60


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€53.60
100 G
€109.00
1 KG
€818.00

About This Item

Empirical Formula (Hill Notation):
H4BK
CAS Number:
Molecular Weight:
53.94
MDL number:
UNSPSC Code:
12352306
EC Index Number:
237-360-5
NACRES:
NA.22

€53.60


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

form

crystals

potency

167 mg/kg LD50, oral (Rat)
230 mg/kg LD50, skin (Rabbit)

reaction suitability

reagent type: reductant

mp

500 °C

density

1.11 g/cm3

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[B-]

InChI

1S/B.K/q-1;+1

InChI key

HHUJLKPGQMFFMS-UHFFFAOYSA-N

Application

  • Hydrolysis for Hydrogen Production: Investigates the catalytic activity of carbon nanotube supported-Ru for hydrogen production via the hydrolysis of potassium borohydride, demonstrating its potential in hydrogen energy systems (Keskin et al., 2022).

Analysis Note

Assay (hypochlorite): ≥ 97.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service