Skip to Content
Merck
All Photos(1)

Key Documents

8.20662

Sigma-Aldrich

2-Hydroxybenzyl alcohol

for synthesis

Synonym(s):

2-Hydroxybenzyl alcohol, Saligenine, Salicyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€45.10
50 G
€70.30
250 G
€307.00

€45.10


0 left in stock

Request a Bulk Order

Select a Size

Change View
10 G
€45.10
50 G
€70.30
250 G
€307.00

About This Item

Linear Formula:
2-(OH)C6H4CH2OH
CAS Number:
Molecular Weight:
124.14
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-960-5
NACRES:
NA.22

€45.10


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

mp

83-86 °C

storage temp.

2-30°C

SMILES string

OCc1c(cccc1)O

InChI

1S/C7H8O2/c8-5-6-3-1-2-4-7(6)9/h1-4,8-9H,5H2

InChI key

CQRYARSYNCAZFO-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 83 °C
Melting range (upper value): ≤ 86 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service