Skip to Content
Merck
All Photos(1)

Key Documents

8.20633

Sigma-Aldrich

n-Hexadecane

for synthesis

Synonym(s):

n-Hexadecane, Cetane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€26.70
250 ML
€117.00
1 L
€315.00
40 KG
€7,650.00

€26.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€26.70
250 ML
€117.00
1 L
€315.00
40 KG
€7,650.00

About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
Molecular Weight:
226.44
MDL number:
UNSPSC Code:
12352001
EC Index Number:
208-878-9
NACRES:
NA.22
grade:
for synthesis
synthesis grade
vapor pressure:
<0.1 hPa ( 20 °C)

€26.70


Check Cart for Availability

Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

dilution

(for synthesis)

kinematic viscosity

<7 cSt(40 °C)

mp

18 °C

transition temp

flash point 135 °C

density

0.77 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCCCC)CCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Application

  • Determination of solvation parameter model compound descriptors by gas chromatography.: This research explores the use of gas chromatography to determine solvation parameters of various compounds, including n-Hexadecane. The study highlights n-Hexadecane′s role in solvation parameter models, which are crucial for understanding solvent-solute interactions in various chemical processes (Poole, 2024).
  • Shear-Responsive Sol-Gel Transition of Phase Change Material Emulsions for an Injectable Thermal Insulation Platform.: This study investigates the phase change properties of n-Hexadecane within emulsions used for thermal insulation. The findings reveal the potential of n-Hexadecane as a key material in advanced thermal management systems, especially for injectable thermal insulation applications (Sun et al., 2023).
  • Microfluidic Fabrication and Thermal Properties of Microencapsulated N-Hexadecane with a Hybrid Polymer Shell for Thermal Energy Storage.: This paper discusses the fabrication of microencapsulated n-Hexadecane using microfluidic techniques. The encapsulation enhances the thermal stability and efficiency of n-Hexadecane as a phase change material, making it suitable for thermal energy storage applications (Yang et al., 2022).
  • Microencapsulated phase change material via Pickering emulsion stabilized by cellulose nanofibrils for thermal energy storage.: The study explores the stabilization of n-Hexadecane within Pickering emulsions using cellulose nanofibrils. The microencapsulation significantly improves the thermal energy storage capabilities of n-Hexadecane, making it an effective component in sustainable energy systems (Bahsi Kaya et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.773 - 0.774
Identity (GC): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the quantity or volume of this product 8.20633 n-Hexadecane?

    1 answer
    1. This product is currently offered as the following as listed on the Product Detail Page:
      8206330005- 5mL
      8206330250- 250mL
      8206331000-1L

      Helpful?

  4. Purity is not mentioned?

    1 answer
    1. This product has an assigned purity specification of ≥99.0%. Lot-specific results are reported on the product's Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service