Skip to Content
Merck
All Photos(1)

Key Documents

8.20599

Sigma-Aldrich

Propyl 3,4,5-trihydroxybenzoate

for synthesis

Synonym(s):

Propyl 3,4,5-trihydroxybenzoate, Propyl gallate, Gallic acid propyl ester, 3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€37.70
100 G
€60.50
500 G
€199.00

€37.70


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€37.70
100 G
€60.50
500 G
€199.00

About This Item

Linear Formula:
3,4,5-(OH)3C6H2COOCH2CH2CH3
CAS Number:
Molecular Weight:
212.20
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-498-2
NACRES:
NA.22

€37.70


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

potency

2100 mg/kg LD50, oral (Rat)

mp

145-148 °C

transition temp

flash point 187 °C

solubility

3.5 g/L

storage temp.

2-30°C

SMILES string

O(CCC)C(=O)c1cc(c(c(c1)O)O)O

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Application

<ul>
<li><strong>Antioxidant Capacity Assessment:</strong> Propyl 3,4,5-trihydroxybenzoate, identified as a potent antioxidant and has been added to foods containing oils and fats to prevent oxidation (Prieto et al., 2014).</li>
</ul>

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 146 °C
Melting range (upper value): ≤ 150 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service