Skip to Content
Merck
All Photos(1)

Key Documents

8.20544

Sigma-Aldrich

1-Dodecanethiol

for synthesis

Synonym(s):

1-Dodecanethiol, Lauryl mercaptan, Dodecyl mercaptan

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€52.60
1 L
€95.20
10 L
€437.00
170 KG
€5,140.00

€52.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€52.60
1 L
€95.20
10 L
€437.00
170 KG
€5,140.00

About This Item

Linear Formula:
CH3(CH2)11SH
CAS Number:
Molecular Weight:
202.40
MDL number:
UNSPSC Code:
12352105
EC Index Number:
203-984-1
NACRES:
NA.22

€52.60


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

0.002 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

230 °C

potency

>2000 mg/kg LD50, oral (Rat)

mp

-9--7 °C

transition temp

flash point 127 °C

density

0.845 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCCCCCCCCCCCC

InChI

1S/C12H26S/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

InChI key

WNAHIZMDSQCWRP-UHFFFAOYSA-N

General description

1-Dodecanethiol is an alkyl thiol that can be used as an organic source of sulfur. It is used in the production of synthetic rubber, nonionic detergent, plastics, and pharmaceuticals.

Application

1-Dodecanethiol (1-DDT) can be used as:      
  • A sulfur source and capping ligand in the synthesis of copper sulfide nanoparticles (Cu2S) which can be applied as electron donor and hole acceptor in photocatalytic reactions.      
  • A coordinating ligand to prepare Cu2ZnSnS4 (CZTS) nanocrystals to control the nucleation and growth of CZTS nanocrystals.

Zinc-doped copper indium sulfide (ZnCuInS2) quantum dots can be functionalized with 1-decanethiol as a ligand to improve the charge mobility and luminance of organic light emitting diodes (OLEDs).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.844 - 0.846
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service