Skip to Content
Merck
All Photos(1)

Key Documents

8.20536

Sigma-Aldrich

N,N′-Diphenyl-1,4-phenylenediamine

for synthesis

Synonym(s):

N,N′-Diphenyl-1,4-phenylenediamine, DPPD

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€108.00
10 KG
€1,280.00

€108.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€108.00
10 KG
€1,280.00

About This Item

Linear Formula:
1,4-(C6H5NH)2C6H4
CAS Number:
Molecular Weight:
260.33
MDL number:
UNSPSC Code:
12352116
EC Index Number:
200-806-4
NACRES:
NA.22

€108.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥95.0% (GC)

form

crystals

potency

2370 mg/kg LD50, oral (Rat)
4500 mg/kg LD50, skin (Rat)

bp

220-225 °C/1.33 hPa

mp

135 °C

density

1.3 g/cm3 at 20 °C

bulk density

560 kg/m3

storage temp.

2-30°C

SMILES string

N(c2ccc(cc2)Nc3ccccc3)c1ccccc1

InChI

1S/C18H16N2/c1-3-7-15(8-4-1)19-17-11-13-18(14-12-17)20-16-9-5-2-6-10-16/h1-14,19-20H

InChI key

UTGQNNCQYDRXCH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service