Skip to Content
Merck
All Photos(2)

Key Documents

8.20499

Sigma-Aldrich

4-(Dimethylamino)pyridine

for synthesis

Synonym(s):

4-(Dimethylamino)pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€24.00
25 G
€41.90
100 G
€123.00
250 G
€239.00
5 KG
€1,280.00
25 KG
€3,840.00

€24.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
€24.00
25 G
€41.90
100 G
€123.00
250 G
€239.00
5 KG
€1,280.00
25 KG
€3,840.00

About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
MDL number:
UNSPSC Code:
12352116
EC Index Number:
214-353-5
NACRES:
NA.22

€24.00


Estimated to ship on12 April 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

420 °C

potency

140 mg/kg LD50, oral (Rat)
90 mg/kg LD50, skin (Rabbit)

pH

11 (20 °C, 60 g/L in H2O)

bp

138-140 °C/24 hPa

mp

110-112 °C

transition temp

flash point 124 °C

solubility

60 g/L

bulk density

520 kg/m3

storage temp.

2-30°C

SMILES string

[N+](=C1C=CNC=C1)(C)C

InChI

1S/C7H10N2/c1-9(2)7-3-5-8-6-4-7/h3-6H,1-2H3/p+1

InChI key

VHYFNPMBLIVWCW-UHFFFAOYSA-O

Application

  • Synthesis and Catalytic Application: Discusses the synthesis of a soluble molecular carrier of sodium hydride activated by DMAP, highlighting its potential in catalysis (Macdonald et al., 2024).
  • Catalyst for Acylation: Describes the use of DMAP hydrochloride as a recyclable catalyst for the acylation of inert alcohols, showing its efficiency and potential for reuse in synthetic processes (Liu et al., 2014).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 110 °C
Melting range (upper value): ≤ 113 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

255.2 °F

Flash Point(C)

124 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service