Skip to Content
Merck
All Photos(1)

Key Documents

8.20446

Sigma-Aldrich

1,3-Dichloro-4-nitrobenzene

for synthesis

Synonym(s):

1,3-Dichloro-4-nitrobenzene, 2,4-Dichloro-1-nitrobenzene, 1-Nitro-2.4-dichlorine benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€356.00

€356.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€356.00

About This Item

Linear Formula:
1,3-(Cl)2C6H3-4-(NO2)
CAS Number:
Molecular Weight:
192.00
MDL number:
UNSPSC Code:
12352101
EC Index Number:
210-248-3
NACRES:
NA.22

€356.00


Estimated to ship on25 May 2025


Request a Bulk Order

vapor pressure

0.013 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

form

crystals

autoignition temp.

500 °C

potency

380 mg/kg LD50, oral (Rat)
920 mg/kg LD50, skin (Rat)

bp

259 °C

mp

28-31 °C

transition temp

flash point 152 °C

density

1.51 g/cm3

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)[N+](=O)[O-]

InChI

1S/C6H3Cl2NO2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H

InChI key

QUIMTLZDMCNYGY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service