Skip to Content
Merck
All Photos(1)

Key Documents

8.20418

Sigma-Aldrich

Ethyl 2,3-dibromopropionate

for synthesis

Synonym(s):

Ethyl 2,3-dibromopropionate, 2,3-Dibromopropionic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€67.50

€67.50


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€67.50

About This Item

Empirical Formula (Hill Notation):
C5H8Br2O2
CAS Number:
Molecular Weight:
259.92
MDL number:
UNSPSC Code:
12352108
EC Index Number:
222-941-8
NACRES:
NA.22

€67.50


Estimated to ship on19 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

240 mg/kg LD50, oral (Rat)
230 mg/kg LD50, skin (Rabbit)

bp

211-214 °C/992 hPa

transition temp

flash point 91 °C

density

1.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrC(CBr)C(=O)OCC

InChI

1S/C5H8Br2O2/c1-2-9-5(8)4(7)3-6/h4H,2-3H2,1H3

InChI key

OENICUBCLXKLJQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.788 - 1.790
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service