Skip to Content
Merck
All Photos(1)

Key Documents

8.20408

Sigma-Aldrich

Dibenzofuran

for synthesis

Synonym(s):

Dibenzofuran, Diphenylene oxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€25.70
250 G
€73.70

€25.70


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
€25.70
250 G
€73.70

About This Item

Empirical Formula (Hill Notation):
C12H8O
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
205-071-3
NACRES:
NA.22

€25.70


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

powder

mp

80-82 °C (External MSDS)

transition temp

flash point 130 °C

density

1.3 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[o]1c2c(c3c1cccc3)cccc2

InChI

1S/C12H8O/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H

InChI key

TXCDCPKCNAJMEE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 79 °C
Melting range (upper value): ≤ 83 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

266.0 °F

Flash Point(C)

130 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service