Skip to Content
Merck
All Photos(1)

Documents

8.20399

Sigma-Aldrich

2-Chloro-N,N-diethylacetamide

for synthesis

Synonym(s):

2-Chloro-N,N-diethylacetamide, Chloroacetic acid diethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CON(C2H5)2
CAS Number:
Molecular Weight:
149.62
MDL number:
UNSPSC Code:
12352111
EC Index Number:
219-019-2

Quality Level

form

liquid

potency

500 mg/kg LD50, oral (Rat)
980 mg/kg LD50, skin (Rat)

bp

114-116 °C/23 hPa

transition temp

flash point 113 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H12ClNO/c1-3-8(4-2)6(9)5-7/h3-5H2,1-2H3

InChI key

CQQUWTMMFMJEFE-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service