Skip to Content
Merck
All Photos(1)

Documents

8.20386

Sigma-Aldrich

Dibutyltin diacetate

for synthesis

Synonym(s):

Dibutyltin diacetate, Diacetoxydibutylstannane, Diacetoxydibutyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2Sn(OOCCH3)2
CAS Number:
Molecular Weight:
351.03
MDL number:
UNSPSC Code:
12352103
EC Index Number:
213-928-8
NACRES:
NA.22

vapor pressure

1.7 hPa ( 25 °C)

Quality Level

autoignition temp.

>200 °C

potency

900-1235 mg/kg LD50, oral (Rat)
2320 mg/kg LD50, skin (Rabbit)

bp

142-145 °C/13 hPa

mp

12 °C

transition temp

flash point 143 °C

solubility

0.006 g/L

density

1.30 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/2C4H9.2C2H3O2.Sn/c2*1-3-4-2;2*1-2(3)4;/h2*1,3-4H2,2H3;2*1H3;

InChI key

QFRLGSUYBLMJKF-UHFFFAOYSA-N

Analysis Note

Assay (ex Sn): ≥ 96.0 %
Density (d 20 °C/ 4 °C): 1.300 - 1.310
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 1 Oral

Target Organs

thymus gland

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.9 °F - Pensky-Martens closed cup

Flash Point(C)

150.5 °C - Pensky-Martens closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service