Skip to Content
Merck
All Photos(2)

Key Documents

8.20359

Sigma-Aldrich

2,4,6-Tris(allyloxy)-1,3,5-triazine

(stabilised) for synthesis

Synonym(s):

2,4,6-Tris(allyloxy)-1,3,5-triazine, Cyanuric acid triallyl ester, Triallyl cyanurate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€56.50
30 KG
€2,190.00

€56.50


Estimated to ship on01 June 2025



Select a Size

Change View
250 G
€56.50
30 KG
€2,190.00

About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
MDL number:
UNSPSC Code:
12352112
EC Index Number:
202-936-7
NACRES:
NA.22

€56.50


Estimated to ship on01 June 2025


vapor pressure

1.3 hPa ( 100 °C)

Quality Level

potency

753 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

149-150 °C/3 hPa (decomposes)

mp

26-27 °C

transition temp

flash point 165-170 °C

density

1.11 g/cm3 at 30 °C

SMILES string

n1c(nc(nc1OCC=C)OCC=C)OCC=C

InChI

1S/C12H15N3O3/c1-4-7-16-10-13-11(17-8-5-2)15-12(14-10)18-9-6-3/h4-6H,1-3,7-9H2

InChI key

BJELTSYBAHKXRW-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 25 °C
Melting range (upper value): ≤ 28 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

347.9 °F

Flash Point(C)

175.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service