Skip to Content
Merck
All Photos(1)

Key Documents

8.20314

Sigma-Aldrich

1-Chloro-2-methylpropane

for synthesis

Synonym(s):

1-Chloro-2-methylpropane, Isobutyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€36.30
250 ML
€65.40

€36.30


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€36.30
250 ML
€65.40

About This Item

Linear Formula:
(CH3)2CHCH2Cl
CAS Number:
Molecular Weight:
92.57
MDL number:
UNSPSC Code:
12352101
EC Index Number:
208-157-9
NACRES:
NA.22

€36.30


Estimated to ship on22 May 2025


Request a Bulk Order

vapor pressure

161 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

395 °C

expl. lim.

2.0-8.8 % (v/v)

bp

69 °C/1013 hPa

mp

-131 °C

transition temp

flash point -17 °C

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCC(C)C

InChI

1S/C4H9Cl/c1-4(2)3-5/h4H,3H2,1-2H3

InChI key

QTBFPMKWQKYFLR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.876 - 0.878
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.9 °F - closed cup

Flash Point(C)

-19.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service