Skip to Content
Merck
All Photos(1)

Key Documents

8.19054

Sigma-Aldrich

1-Hydroxy-2-pyridinthione sodium salt

(40% solution in water)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€87.50

€87.50


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€87.50

About This Item

UNSPSC Code:
12352107
NACRES:
NA.22

€87.50


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

form

liquid

pH

8.5-10.5 (20 °C in H2O)

bp

100 °C/1013 hPa

density

1.22 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (iodometric): 38.0 - 42.0 %
Density (d 20 °C/ 4 °C): 1.190 - 1.240

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Nervous system

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service