Skip to Content
Merck
All Photos(1)

Key Documents

8.18816

Sigma-Aldrich

3-(Chloropropyl)-trimethoxysilane

for synthesis

Synonym(s):

3-(Chloropropyl)-trimethoxysilane, CPTMO

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€108.00

€108.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€108.00

About This Item

Linear Formula:
Cl(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
198.72
MDL number:
UNSPSC Code:
12352103
EC Index Number:
219-787-9
NACRES:
NA.22

€108.00


Estimated to ship on23 May 2025


Request a Bulk Order

vapor pressure

<5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

220 °C

potency

>2000 mg/kg LD50, oral (Rat)
3062 mg/kg LD50, skin (Rabbit)

expl. lim.

5.5-44 % (v/v)

bp

195-197 °C/1013 hPa

mp

-50 °C

transition temp

flash point 78 °C

density

1.082 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](OC)(OC)(OC)CCCCl

InChI

1S/C6H15ClO3Si/c1-8-11(9-2,10-3)6-4-5-7/h4-6H2,1-3H3

InChI key

OXYZDRAJMHGSMW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.081 - 1.085
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.1 °F - closed cup

Flash Point(C)

71.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service