Skip to Content
Merck
All Photos(1)

Key Documents

8.18793

Sigma-Aldrich

2-Nonanol

for synthesis

Synonym(s):

2-Nonanol, Heptyl methyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€48.20
100 ML
€134.00

€48.20


0 left in stock


Select a Size

Change View
25 ML
€48.20
100 ML
€134.00

About This Item

Linear Formula:
CH3(CH2)6CH(OH)CH3
CAS Number:
Molecular Weight:
144.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
211-065-1
NACRES:
NA.22

€48.20


0 left in stock

Quality Level

Assay

≥98.0% (GC)

mp

-35 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CCCCCCC)C

InChI

1S/C9H20O/c1-3-4-5-6-7-8-9(2)10/h9-10H,3-8H2,1-2H3

InChI key

NGDNVOAEIVQRFH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.821 - 0.823
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service