Skip to Content
Merck
All Photos(1)

Key Documents

8.18758

Sigma-Aldrich

Dimethyl dicarbonate

for synthesis

Synonym(s):

Dimethyl dicarbonate, Dimethyl pyrocarbonate, Pyrocarbonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€297.00

€297.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€297.00

About This Item

Linear Formula:
(CH3OCO)2O
CAS Number:
Molecular Weight:
134.09
MDL number:
UNSPSC Code:
12352108
EC Index Number:
224-859-8
NACRES:
NA.22

€297.00


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

0.7 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (acidimetric)

form

liquid

autoignition temp.

465 °C

potency

335 mg/kg LD50, oral (Rat)
>1250 mg/kg LD50, skin (Rat)

expl. lim.

3-29.9 % (v/v)

bp

172 °C/1013 hPa (decomposes)

mp

15-17 °C

transition temp

flash point 85 °C

solubility

35 g/L (decomposition)

density

1.25 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)OC(=O)OC

InChI

1S/C4H6O5/c1-7-3(5)9-4(6)8-2/h1-2H3

InChI key

GZDFHIJNHHMENY-UHFFFAOYSA-N

Application

  • Enhancing microbial control in beverage production: A study evaluated the combined effect of high pressure processing and dimethyl dicarbonate (DMDC) for inactivating foodborne pathogens in apple juice, demonstrating DMDC′s efficacy as a microbial control agent in beverage preservation (Petrus et al., 2020).
  • Alternative to heat treatment in juice fermentation: Research explored the effects of dimethyl dicarbonate (DMDC) on the fermentation of litchi juice by Lactobacillus casei, presenting DMDC as a viable non-thermal preservation method that could replace traditional heat treatments in the food industry (Yu et al., 2014).

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Im confused as I look at your compound dimethylcarbonate no 818758 its says Assay > 97 % in the specification but beneath in the text on the website it says Assay >99%? Whats the purity and when did you change from 99 to 97 % if its the correct number

    1 answer
    1. The minimum purity specification for this product was changed from 99% to 97% in November of 2022. The specification was updated in the product properties section but not the Analysis Note. This content has now been corrected. Thank you for bringing this to our attention.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service