Skip to Content
Merck
All Photos(1)

Key Documents

8.18739

Sigma-Aldrich

3-Methylphenyl isothiocyanate

for synthesis

Synonym(s):

3-Methylphenyl isothiocyanate, 3-Tolyl isothiocyanate, Isothiocyanic acid 3-methylphenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(CH3)C6H4NCS
CAS Number:
Molecular Weight:
149.21
MDL number:
UNSPSC Code:
12352118
EC Index Number:
210-677-6

Quality Level

Assay

≥99.0% (GC)

form

liquid

bp

243 °C/1013 hPa

transition temp

flash point 106 °C

density

1.10 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H7NS/c1-7-3-2-4-8(5-7)9-6-10/h2-5H,1H3

InChI key

BDPQUWSFKCFOST-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.098 - 1.102
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service