Skip to Content
Merck
All Photos(1)

Key Documents

8.18737

Sigma-Aldrich

Chlorotrimethylsilane

for synthesis

Synonym(s):

Chlorotrimethylsilane, Trimethylchlorosilane, Trimethylsilyl chloride, TMCS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€35.80
500 ML
€83.70
23 KG
€922.00

€35.80


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€35.80
500 ML
€83.70
23 KG
€922.00

About This Item

Linear Formula:
(CH3)3SiCl
CAS Number:
Molecular Weight:
108.64
MDL number:
UNSPSC Code:
12352103
EC Index Number:
200-900-5
NACRES:
NA.22

€35.80


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

250 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

420 °C

potency

4868 mg/kg LD50, oral (Rat)
1530 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-46 % (v/v)

bp

57 °C/1013 hPa

mp

-58 °C

transition temp

flash point -20 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(C)(C)C

InChI

1S/C3H9ClSi/c1-5(2,3)4/h1-3H3

InChI key

IJOOHPMOJXWVHK-UHFFFAOYSA-N

Application

  • Synthesis of bis ferrocene derivatives: Development of new ferrocenyl multi-functional silyl ether compounds using chlorotrimethylsilane, highlighting advances in organosilicon chemistry (R Teimuri-Mofrad et al., 2016).
  • Pharmacologically active derivatives: Use of chlorotrimethylsilane as a catalyst in the synthesis of pharmacologically active bis (indolyl) and tris (indolyl) derivatives, demonstrating its efficiency in electrophilic substitution reactions (NG Singh et al., 2014).
  • Silicone resin synthesis: Exploration of the hydrolysis-condensation reaction of tetraethoxysilane and chlorotrimethylsilane to create silicone resins, showing the material′s enhanced properties for industrial applications (CFJ Kuo et al., 2014).
  • Deoxygenation of unsymmetrical dicarbonyl compounds: Innovative use of chlorotrimethylsilane and sodium iodide for the selective reduction of 1,2-diketones, showcasing a novel method in organic synthesis (LZ Yuan et al., 2016).
  • Enhanced activity and durability of catalysts: Investigation into Fe3+ and chlorotrimethylsilane modified NaY catalysts for the acetalization of glycerol to solketal, improving catalytic performance in chemical processes (Y Li et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.858 - 0.861
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service