Skip to Content
Merck
All Photos(1)

Key Documents

8.18715

Sigma-Aldrich

Paraformaldehyde

for synthesis

Synonym(s):

Paraformaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€33.30
1 KG
€51.90
25 KG
€454.00

€33.30


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€33.30
1 KG
€51.90
25 KG
€454.00

About This Item

Linear Formula:
(CH2O)n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.22
grade:
for synthesis
technique(s):
immunofluorescence: suitable
vapor pressure:
1.93 hPa ( 25 °C)

€33.30


Estimated to ship on23 May 2025


Request a Bulk Order

grade

for synthesis

Quality Level

vapor pressure

1.93 hPa ( 25 °C)

Assay

≥95% (acidimetric)

form

solid

autoignition temp.

300 °C

potency

592 mg/kg LD50, oral (Rat)

expl. lim.

7-73 % (v/v)

technique(s)

immunofluorescence: suitable

pH

5.5 (20 °C in H2O, saturated solution)

mp

100-130 °C

density

1.4 g/cm3 at 20 °C

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Application

  • Flame Retardants: Examines the use of paraformaldehyde in improving the performance of flame retardants without added antioxidants (P Sun, et al., 2024).

Linkage

Replaces: PX0055-3

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service