Skip to Content
Merck
All Photos(1)

Key Documents

8.18631

Sigma-Aldrich

Zinc bromide

anhydrous for synthesis

Synonym(s):

Zinc bromide, Zinc dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€28.90
250 G
€89.80
1 KG
€249.00

€28.90


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€28.90
250 G
€89.80
1 KG
€249.00

About This Item

Empirical Formula (Hill Notation):
Br2Zn
CAS Number:
Molecular Weight:
225.20
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-718-4
NACRES:
NA.22

€28.90


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

form

crystals

pH

4 (20 °C in H2O, saturated solution)

bp

697 °C/1013 mbar

mp

394 °C

solubility

4470 g/L

density

4.2 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Zn](Br)Br

InChI

1S/2BrH.Zn/h2*1H;/q;;+2/p-2

InChI key

VNDYJBBGRKZCSX-UHFFFAOYSA-L

Application

Zinc bromide can be used as:
  • A catalyst to synthesize β-hydroxy ketones by stereoselective semipinacol rearrangement of α-hydroxy epoxides.[1]
  • A safe brominating source for the oxidative bromination of alkenes and alkynes in combination with lead tetraacetate.[2]

It can also be used to prepare silica-supported zinc bromide (ZnBr2/SiO2) composite, which is used as a catalyst to synthesize:
  • Ynone derivatives via cross-coupling reaction of acid chlorides with terminal alkynes.[3]
  • 4,5-disubstituted-1,2,3-(NH)-triazole derivatives by one-pot cross-coupling and 1,3-dipolar cycloaddition reaction of acid chlorides, terminal alkynes, and sodium azide.[3]

Analysis Note

Assay (argentometric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service