Skip to Content
Merck
All Photos(1)

Key Documents

8.18627

Sigma-Aldrich

Linalool

for synthesis

Synonym(s):

Linalool, 3,7-Dimethyl-1,6-octadien-3-ol

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€24.90
250 ML
€59.60

€24.90


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€24.90
250 ML
€59.60

About This Item

Linear Formula:
CH3C(CH3)=CH(CH2)2C(OH)(CH3)CH=CH2
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-134-4
NACRES:
NA.22

€24.90


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

0.10 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

235 °C

potency

2790 mg/kg LD50, oral (Rat)
5610 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-5.2 % (v/v)

pH

4.5 (25 °C, 1.45 g/L in H2O)

transition temp

flash point 79 °C

solubility

1.45 g/L

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CCC=C(C)C)(C)C=C

InChI

1S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11H,1,6,8H2,2-4H3

InChI key

CDOSHBSSFJOMGT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.860 - 0.862
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

171.0 °F - Pensky-Martens closed cup

Flash Point(C)

77.2 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service