Skip to Content
Merck
All Photos(1)

Key Documents

8.18601

Sigma-Aldrich

di-Sodium succinate

anhydrous for synthesis

Synonym(s):

di-Sodium succinate, Succinic acid disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€44.00
500 G
€125.00
5 KG
€314.00

€44.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€44.00
500 G
€125.00
5 KG
€314.00

About This Item

Empirical Formula (Hill Notation):
C4H4Na2O4
CAS Number:
Molecular Weight:
162.05
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-778-7
NACRES:
NA.22

€44.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

form

powder

pH

7-9 (20 °C, 100 g/L in H2O)

mp

>300 °C

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]C(=O)CCC(=O)[O-]

InChI

1S/C4H6O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H2,(H,5,6)(H,7,8);;/q;2*+1/p-2

InChI key

ZDQYSKICYIVCPN-UHFFFAOYSA-L

Analysis Note

Assay (HClO₄): ≥ 96.0 %
Loss on drying: ≤ 3.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service