Skip to Content
Merck
All Photos(1)

Key Documents

8.18587

Sigma-Aldrich

2,5-Dimethyl-3-hexyne-2,5-diol

for synthesis

Synonym(s):

2,5-Dimethyl-3-hexyne-2,5-diol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€71.60

€71.60


Estimated to ship on21 May 2025



Select a Size

Change View
100 G
€71.60

About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
MDL number:
UNSPSC Code:
12352104
EC Index Number:
205-533-4
NACRES:
NA.22

€71.60


Estimated to ship on21 May 2025


vapor pressure

0.025 hPa ( 40.3 °C)

Quality Level

Assay

≥99.0% (GC)

bp

205 °C/1013 hPa

mp

93-95 °C

solubility

240 g/L

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(C)(C)C#CC(O)(C)C

InChI

1S/C8H14O2/c1-7(2,9)5-6-8(3,4)10/h9-10H,1-4H3

InChI key

IHJUECRFYCQBMW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 93 °C
Melting range (upper value): ≤ 95 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service