8.18582
N-Formylmorpholine
for synthesis
Synonym(s):
N-Formylmorpholine, 4-Morpholinecarboxaldehyde
Select a Size
€36.80
Select a Size
About This Item
€36.80
Recommended Products
vapor pressure
0.03 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
autoignition temp.
345 °C (DIN 51794)
potency
>7360 mg/kg LD50, oral (Rat)
>18400 mg/kg LD50, skin (Rabbit)
expl. lim.
1.2-8.2 % (v/v)
pH
10 (20 °C, 250 g/L in H2O)
bp
240 °C/1013 hPa
mp
23 °C
transition temp
flash point 118 °C (DIN 51758)
density
1.14 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
N1(CCOCC1)C=O
InChI
1S/C5H9NO2/c7-5-6-1-3-8-4-2-6/h5H,1-4H2
InChI key
LCEDQNDDFOCWGG-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.149 - 1.151
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
244.4 °F - DIN 51758
Flash Point(C)
118 °C - DIN 51758
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service