Skip to Content
Merck
All Photos(1)

Key Documents

8.18580

Sigma-Aldrich

Methoxymethyl-triphenylphosphonium chloride

for synthesis

Synonym(s):

Methoxymethyl-triphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€92.90

€92.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€92.90

About This Item

Linear Formula:
CH3OCH2P(C6H5)3Cl
CAS Number:
Molecular Weight:
342.80
MDL number:
UNSPSC Code:
12352107
EC Index Number:
223-664-5
NACRES:
NA.22

€92.90


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

powder

potency

1440 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: C-C Bond Formation

pH

2.2 (20 °C, 1100 g/L in H2O)

mp

202 °C

transition temp

flash point >250 °C

solubility

soluble >1100 g/L (decomposition)

bulk density

450‑500 kg/m3

storage temp.

2-30°C

SMILES string

[P+](COC)(c3ccccc3)(c2ccccc2)c1ccccc1.[Cl-]

InChI

1S/C20H20OP.ClH/c1-21-17-22(18-11-5-2-6-12-18,19-13-7-3-8-14-19)20-15-9-4-10-16-20;/h2-16H,17H2,1H3;1H/q+1;/p-1

InChI key

SJFNDMHZXCUXSA-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

482.0 °F

Flash Point(C)

> 250 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service