8.18555
2,3-Pentanedione
for synthesis
Synonym(s):
2,3-Pentanedione
Select a Size
€36.80
Estimated to ship on29 May 2025
Select a Size
About This Item
€36.80
Estimated to ship on29 May 2025
vapor pressure
28.5 hPa ( 20 °C)
Quality Level
Assay
≥97.5% (GC)
autoignition temp.
265 °C
potency
3000 mg/kg LD50, oral (Rat)
>2500 mg/kg LD50, skin (Rabbit)
expl. lim.
1.8-10.9 % (v/v)
pH
4 ( in H2O)
mp
-52 °C
transition temp
flash point 18 °C
solubility
60 g/L
density
0.96 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O=C(CC)C(=O)C
InChI
1S/C5H8O2/c1-3-5(7)4(2)6/h3H2,1-2H3
InChI key
TZMFJUDUGYTVRY-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 0.958 - 0.966
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Sens. 1B - STOT RE 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
66.2 °F - open cup
Flash Point(C)
19 °C - open cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service