Skip to Content
Merck
All Photos(1)

Key Documents

8.18401

Sigma-Aldrich

Potassium hydrogen monopersulfate

for synthesis

Synonym(s):

Potassium hydrogen monopersulfate, PMPS, KMPS

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€32.00
500 G
€50.30

€32.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
100 G
€32.00
500 G
€50.30

About This Item

Empirical Formula (Hill Notation):
H3K5O18S4
CAS Number:
Molecular Weight:
614.76
MDL number:
UNSPSC Code:
12352302
EC Index Number:
274-778-7
NACRES:
NA.22

€32.00


0 left in stock

Request a Bulk Order

Quality Level

form

powder

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

pH

2-3 (20 °C, 10 g/L in H2O)

solubility

soluble 250-300 g/L

density

2.35 g/cm3 at 20 °C

Analysis Note

Assay (iodometric): 43.0 - 47.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service