Skip to Content
Merck
All Photos(2)

Key Documents

8.18256

Sigma-Aldrich

Cyclohexanecarboxylic acid chloride

for synthesis

Synonym(s):

Cyclohexanecarboxylic acid chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€62.40

€62.40


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€62.40

About This Item

Empirical Formula (Hill Notation):
C7H11ClO
CAS Number:
Molecular Weight:
146.61
MDL number:
UNSPSC Code:
12352106
EC Index Number:
220-322-7
NACRES:
NA.22

€62.40


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

form

liquid

potency

965 mg/kg LD50, oral (Rat)

bp

180-181 °C/1013 hPa

transition temp

flash point 66 °C

density

1.08 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(=O)C1CCCCC1

InChI

1S/C7H11ClO/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2

InChI key

RVOJTCZRIKWHDX-UHFFFAOYSA-N

Analysis Note

Assay (morpholine method): ≥ 97.0 %
Density (d 20 °C/ 4 °C): 1.081 - 1.087
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

150.8 °F

Flash Point(C)

66 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service