Skip to Content
Merck
All Photos(1)

Key Documents

8.18248

Sigma-Aldrich

Dichlorophenylphosphine oxide

for synthesis

Synonym(s):

Dichlorophenylphosphine oxide, Phenylphosphonic acid dichloride, Benzenephosphonic dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€131.00

€131.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€131.00

About This Item

Linear Formula:
C6H5P(O)Cl2
CAS Number:
Molecular Weight:
194.98
MDL number:
UNSPSC Code:
12352106
EC Index Number:
212-534-3
NACRES:
NA.22

€131.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

681 mg/kg LD50, oral (Rat)

mp

1 °C

transition temp

flash point 204 °C

density

1.40 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[P](=O)(Cl)(Cl)c1ccccc1

InChI

1S/C6H5Cl2OP/c7-10(8,9)6-4-2-1-3-5-6/h1-5H

InChI key

IBDMRHDXAQZJAP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.396 - 1.400
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

399.2 °F

Flash Point(C)

204 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service