Skip to Content
Merck
All Photos(1)

Key Documents

8.18246

Sigma-Aldrich

Tetraethylammonium tetrafluoroborate

for synthesis

Synonym(s):

Tetraethylammonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4NBF4
CAS Number:
Molecular Weight:
217.06
MDL number:
UNSPSC Code:
12352107
EC Index Number:
207-055-1
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
crystals
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

Assay

≥97.0% (acidimetric)

form

crystals

mp

365-368 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[B]([F-])(F)(F)F.[N+](CC)(CC)(CC)CC

InChI

1S/C8H20N.BF4/c1-5-9(6-2,7-3)8-4;2-1(3,4)5/h5-8H2,1-4H3;/q+1;-1

InChI key

XJRAKUDXACGCHA-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Assay (ex H₃BO₃): ≥ 97.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service