Skip to Content
Merck
All Photos(1)

Key Documents

8.18090

Sigma-Aldrich

Chromium(III) chloride

anhydrous for synthesis

Synonym(s):

Chromium(III) chloride, Chromium trichloride

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€35.30
25 G
€83.70

€35.30


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
2 G
€35.30
25 G
€83.70

About This Item

Empirical Formula (Hill Notation):
Cl3Cr
CAS Number:
Molecular Weight:
158.36
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-038-3
NACRES:
NA.22
grade:
for synthesis
form:
powder

€35.30


Estimated to ship on22 May 2025


Request a Bulk Order

grade

for synthesis

Quality Level

form

powder

potency

440 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: reductant

pH

2.4-2.6 (20 °C, 50 - 53 g/L in H2O)

mp

1152 °C

density

2.87 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Cr+3].[Cl-].[Cl-].[Cl-].O.O.O.O.O.O

InChI

1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

LJAOOBNHPFKCDR-UHFFFAOYSA-K

Analysis Note

Assay (iodometric): ≥ 98.0 %
Loss on drying (105 °C, 2 h): ≤ 1.0 %
Identity (ICP-OES): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service