Skip to Content
Merck
All Photos(1)

Key Documents

8.18084

Sigma-Aldrich

3-Hydroxypropionitrile

for synthesis

Synonym(s):

3-Hydroxypropionitrile, ß-Cyanoethanol, Ethylene cyanohydrin

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€44.00
500 ML
€85.10
25 KG
€2,120.00

€44.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€44.00
500 ML
€85.10
25 KG
€2,120.00

About This Item

Empirical Formula (Hill Notation):
C3H5NO
CAS Number:
Molecular Weight:
71.08
MDL number:
UNSPSC Code:
12352117
EC Index Number:
203-704-8
NACRES:
NA.22

€44.00


Estimated to ship on16 May 2025


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

3200 mg/kg LD50, oral (Rat)
5250 mg/kg LD50, skin (Rabbit)

pH

3.0-4.5 (20 °C in H2O, undiluted)

bp

228 °C/1013 hPa

mp

-46 °C

transition temp

flash point 128 °C (DIN 51758)

solubility

1000 g/L

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N#CCCO

InChI

1S/C3H5NO/c4-2-1-3-5/h5H,1,3H2

InChI key

WSGYTJNNHPZFKR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.045 - 1.047
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service