Skip to Content
Merck
All Photos(2)

Key Documents

8.17042

SAFC

Saccharin sodium dihydrate

EMPROVE® ESSENTIAL, Ph. Eur., USP

Pharma Manufacturing

Synonym(s):

Saccharin sodium dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€231.00
5 KG
€590.00
25 KG
Inquire

€231.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€231.00
5 KG
€590.00
25 KG
Inquire

About This Item

Empirical Formula (Hill Notation):
C7H4NNaO3S · 2H2O
CAS Number:
Molecular Weight:
241.20
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-886-1

€231.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Agency

Ph. Eur.
USP

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

pH

6.0-7.5 (20 °C, 100 g/L in H2O)

transition temp

flash point >200 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[Na+].[S]1(=NC(=O)c2c1cccc2)(=O)[O-].O.O

InChI

1S/C7H5NO3S.Na.2H2O/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;;;/h1-4H,(H,8,9,10,11);;2*1H2/q;+1;;/p-1

InChI key

NAUHOSQQKJTUAX-UHFFFAOYSA-M

Related Categories

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Saccharine sodium is a commonly used ingredient in food and pharmaceutics. In pharmaceutical applications, saccharine sodium has been used in a wide range of different dosage forms as a taste modifier.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service