Skip to Content
Merck
All Photos(1)

Key Documents

8.14779

Sigma-Aldrich

4-Methylmorpholine 4-oxide

(50% solution in water) for synthesis

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€41.20
1 L
€97.10

€41.20


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€41.20
1 L
€97.10

About This Item

UNSPSC Code:
12352119
NACRES:
NA.22

€41.20


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

form

liquid

potency

>9200 mg/kg LD50, oral (Rat)
>8000 mg/kg LD50, skin (Rabbit)

bp

100 °C

mp

-70 °C

density

1.14 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (ex N): 49.0 - 51.0 %
Density (d 20 °C/ 4 °C): 1.105 - 1.115
Water (K. F.): 49.0 - 51.0 %

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service