Skip to Content
Merck
All Photos(2)

Key Documents

8.14667

Sigma-Aldrich

(2-Bromoethyl) ethyl ether

for synthesis

Synonym(s):

(2-Bromoethyl) ethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€106.00

€106.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€106.00

About This Item

Linear Formula:
Br-CH2CH2-O-CH2CH3
CAS Number:
Molecular Weight:
153.02
MDL number:
UNSPSC Code:
12352112
EC Index Number:
209-763-6
NACRES:
NA.22

€106.00


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥95.0% (GC)

form

liquid

bp

125-127 °C/1013 hPa

transition temp

flash point 21 °C

density

1.35 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCCOCC

InChI

1S/C4H9BrO/c1-2-6-4-3-5/h2-4H2,1H3

InChI key

MMYKTRPLXXWLBC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.345 - 1.355
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service