Skip to Content
Merck
All Photos(1)

Key Documents

8.14658

Sigma-Aldrich

Copper(II) bromide

for synthesis

Synonym(s):

Copper(II) bromide, Copper dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€31.90
250 G
€76.20

€31.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€31.90
250 G
€76.20

About This Item

Empirical Formula (Hill Notation):
Br2Cu
CAS Number:
Molecular Weight:
223.35
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-167-2
NACRES:
NA.22

€31.90


Check Cart for Availability

Request a Bulk Order

Quality Level

form

crystals

potency

536 mg/kg LD50, oral (Rat)

bp

900 °C/1013 hPa

mp

498 °C

solubility

1200 g/L

density

4.71 g/cm3 at 20 °C

bulk density

2000 kg/m3

storage temp.

2-30°C

SMILES string

[Cu](Br)Br

InChI

1S/2BrH.Cu/h2*1H;/q;;+2/p-2

InChI key

QTMDXZNDVAMKGV-UHFFFAOYSA-L

Analysis Note

Assay (iodometric): ≥ 98.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service