Skip to Content
Merck
All Photos(1)

Key Documents

8.14632

Sigma-Aldrich

L-Glutamic acid hydrochloride

for synthesis

Synonym(s):

L-Glutamic acid hydrochloride, 2-Aminoglutaric acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€37.20

€37.20


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€37.20

About This Item

Empirical Formula (Hill Notation):
C5H9NO4 · xHCl
CAS Number:
Molecular Weight:
147.13 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-315-9
NACRES:
NA.22

€37.20


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

form

powder

mp

191-193 °C (decomposes)

density

1.525 g/cm3

bulk density

630 kg/m3

storage temp.

2-30°C

SMILES string

Cl.NC(CCC(=O)O)C(=O)O

InChI

1S/C5H9NO4.ClH/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);1H

InChI key

RPAJSBKBKSSMLJ-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (argentometric): ≥ 98.0 %
Spec. rotation [α²0/D (c=10 in HCl 2 mol/l): +24.5 - +26.0 °
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service