Skip to Content
Merck
All Photos(1)

Key Documents

8.14578

Sigma-Aldrich

2,3-Naphthalenediol

for synthesis

Synonym(s):

2,3-Naphthalenediol, 2,3-Dihydroxynaphthalene

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€81.20

€81.20


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€81.20

About This Item

Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-156-7
NACRES:
NA.22

€81.20


Estimated to ship on18 May 2025


Request a Bulk Order

vapor pressure

0.8 hPa ( 149 °C)

Quality Level

Assay

≥97.0% (HPLC)

form

powder

autoignition temp.

>330 °C

potency

970 mg/kg LD50, oral (Rat)

pH

6 (20 °C, 1 g/L in H2O)

mp

161-164 °C

transition temp

flash point 175 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

Oc1cc2c(cc1O)cccc2

InChI

1S/C10H8O2/c11-9-5-7-3-1-2-4-8(7)6-10(9)12/h1-6,11-12H

InChI key

JRNGUTKWMSBIBF-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 161 °C
Melting range (upper value): ≤ 164 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

347.0 °F

Flash Point(C)

175 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service