Skip to Content
Merck
All Photos(1)

Key Documents

8.14538

Sigma-Aldrich

1,2-Phenylenediamine

for synthesis

Synonym(s):

1,2-Phenylenediamine, 1,2-Diaminobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€32.30
250 G
€54.80
1 KG
€155.00

€32.30


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€32.30
250 G
€54.80
1 KG
€155.00

About This Item

Linear Formula:
1,2-(NH2)2C6H4
CAS Number:
Molecular Weight:
108.14
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-430-6
NACRES:
NA.22

€32.30


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

0.013 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

540 °C

expl. lim.

1.5 % (v/v)

pH

7-8 (20 °C, 50 g/L in H2O)

bp

257 °C/1013 hPa

mp

99-103 °C

transition temp

flash point 110 °C

solubility

54 g/L

density

1.14 g/cm3 at 20 °C

bulk density

530 kg/m3

storage temp.

2-30°C

SMILES string

Nc1c(cccc1)N

InChI

1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2

InChI key

GEYOCULIXLDCMW-UHFFFAOYSA-N

Application

1,2-Phenylenediamine for synthesis. CAS 95-54-5, chemical formula 1,2-(NH₂)₂C₆H₄.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 99 °C
Melting range (upper value): ≤ 103 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service