Skip to Content
Merck
All Photos(1)

Key Documents

8.14513

Sigma-Aldrich

3-Chloro-1-methoxy-2-propanol

for synthesis

Synonym(s):

3-Chloro-1-methoxy-2-propanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€70.10

€70.10


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€70.10

About This Item

Empirical Formula (Hill Notation):
C4H9ClO2
CAS Number:
Molecular Weight:
124.57
MDL number:
UNSPSC Code:
12352104
EC Index Number:
223-982-4
NACRES:
NA.22

€70.10


Estimated to ship on15 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

340 mg/kg LD50, oral (Rat)

pH

5.6 (50 g/L in H2O)

transition temp

flash point 72 °C

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClCC(O)COC

InChI

1S/C4H9ClO2/c1-7-3-4(6)2-5/h4,6H,2-3H2,1H3

InChI key

FOLYKNXDLNPWGC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.162 - 1.165
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service