Skip to Content
Merck
All Photos(1)

Key Documents

8.14505

Sigma-Aldrich

1,2,4-Trimethylbenzene

for synthesis

Synonym(s):

1,2,4-Trimethylbenzene, Pseudocumol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€27.00
500 ML
€44.10

€27.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€27.00
500 ML
€44.10

About This Item

Empirical Formula (Hill Notation):
C9H12
CAS Number:
Molecular Weight:
120.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-436-9
NACRES:
NA.22

€27.00


Check Cart for Availability

Request a Bulk Order

vapor pressure

2.8 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

485 °C

potency

5000 mg/kg LD50, oral (Rat)

expl. lim.

0.8-7 % (v/v)

bp

168 °C/1013 hPa

mp

-44 °C

transition temp

flash point 48 °C

solubility

0.057 g/L

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Cc1c(ccc(c1)C)C

InChI

1S/C9H12/c1-7-4-5-8(2)9(3)6-7/h4-6H,1-3H3

InChI key

GWHJZXXIDMPWGX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.874 - 0.876
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service