Skip to Content
Merck
All Photos(1)

Key Documents

8.14495

Sigma-Aldrich

3-Amino-1H-1,2,4-triazole

for synthesis

Synonym(s):

3-Amino-1H-1,2,4-triazole, Amitrol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€48.10
500 G
€172.00

€48.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€48.10
500 G
€172.00

About This Item

Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08
MDL number:
UNSPSC Code:
12352116
EC Index Number:
200-521-5
NACRES:
NA.22

€48.10


Check Cart for Availability

Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

>225 °C

potency

1100 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rat)

pH

6.4-7.4 (20 °C, 100 g/L in H2O)

mp

149-152 °C

solubility

280 g/L

density

1.138 g/cm3 at 20 °C

bulk density

650 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1c(nnc1)N

InChI

1S/C2H4N4/c3-2-4-1-5-6-2/h1H,(H3,3,4,5,6)

InChI key

KLSJWNVTNUYHDU-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Melting range (lower value): ≥ 151 °C
Melting range (upper value): ≤ 157 °C
Identity (IR): passes test

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service