Skip to Content
Merck
All Photos(2)

Key Documents

8.14473

Sigma-Aldrich

(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone

for synthesis

Synonym(s):

(S)-(-)-2-(N-Benzylprolyl)aminobenzophenone, BPB

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€134.00

€134.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 G
€134.00

About This Item

Empirical Formula (Hill Notation):
C25H24N2O2
CAS Number:
Molecular Weight:
384.47
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

€134.00


Estimated to ship on28 May 2025


Quality Level

mp

97-99 °C

storage temp.

2-30°C

SMILES string

[N+H]2([C@@H](CCC2)C(=O)Nc3c(cccc3)C(=O)c4ccccc4)Cc1ccccc1

InChI

1S/C25H24N2O2/c28-24(20-12-5-2-6-13-20)21-14-7-8-15-22(21)26-25(29)23-16-9-17-27(23)18-19-10-3-1-4-11-19/h1-8,10-15,23H,9,16-18H2,(H,26,29)/p+1/t23-/m0/s1

InChI key

IPSABLMEYFYEHS-QHCPKHFHSA-O

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Melting range (lower value): ≥ 100 °C
Melting range (upper value): ≤ 103 °C
Spec. rotation [α²0/D (c=1 in methanol): -136.0 - -134.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service