Skip to Content
Merck
All Photos(2)

Key Documents

8.14393

Sigma-Aldrich

Glutardialdehyde

(50% solution in water) for synthesis

Synonym(s):

Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€30.40
1 L
€74.60
50 KG
€1,050.00

€30.40


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€30.40
1 L
€74.60
50 KG
€1,050.00

About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
100.7 °C/1013 hPa
vapor pressure:
22 hPa ( 20 °C)

€30.40


Estimated to ship on01 June 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

22 hPa ( 20 °C)

form

liquid

autoignition temp.

395 °C

concentration

47.0-53.0% (hydrogensulfite method)

dilution

(for synthesis)

pH

3.1-4.5 (20 °C in H2O)

kinematic viscosity

12.75 cSt(25 °C)

bp

100.7 °C/1013 hPa

mp

-33 °C

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (hydrogensulfit methode): 47.0 - 53.0 %
Density (d 20 °C/ 4 °C): 1.125 - 1.130

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service